Learn More
MSC iGaging Bore Gage Set, 0.7-6 in, 0.00005 in, iP54 Rated
Catalog No. 5000513475
Encompass
This extended range digital bore gage set is designed for close tolerance inspection of bores. It accurately measures size, roundness, ovality, and taper. The absolute electronic bore gage includes an indicator fully protected by a rugged cover and comes with anvils, spacers, and sub-anvils, all furnished in a fitted case.
  • iP54 protective rating for moisture and dust resistance
  • Wide measuring range of 0.7-6 inches
  • High resolution of 0.00005 inches
  • SPC/USB data output capability
  • Three interchangeable measuring heads for versatility
  • Multiple sets of anvils with durable carbide points
  • Includes a 2 inch extender for deeper measurements
  • Three spacers/washers for fine adjustments
Catalog No. 50-005-13475 Supplier MSC Supplier No. 17621376
This extended range digital bore gage set is designed for close tolerance inspection of bores. It accurately measures size, roundness, ovality, and taper. The absolute electronic bore gage includes an indicator fully protected by a rugged cover and comes with anvils, spacers, and sub-anvils, all furnished in a fitted case.
  • iP54 protective rating for moisture and dust resistance
  • Wide measuring range of 0.7-6 inches
  • High resolution of 0.00005 inches
  • SPC/USB data output capability
  • Three interchangeable measuring heads for versatility
  • Multiple sets of anvils with durable carbide points
  • Includes a 2 inch extender for deeper measurements
  • Three spacers/washers for fine adjustments