Learn More
MSC iGaging Coaxial Dial Indicator Kit, 0.0005 inch Graduation, 0.0005 inch Locating Accuracy
Catalog No. 5000527177
Encompass
This coaxial centering indicator is designed to quickly center workpieces or fixtures on a mill. It operates at any angle from horizontal to vertical without changes, and can be used for both internal and external measurements by simply changing the feeler.
  • Quickly centers workpieces or fixtures on a mill.
  • Includes 6 feelers: 4 inch, 8 inch, 12 inch straight; 4 inch, 8 inch, 12 inch curved.
  • Operates at any angle from horizontal to vertical without requiring adjustments.
  • Adaptable for internal and external measurements by switching feelers.
  • Comes supplied in a fitted wood case.
  • Set includes indicator body, restraining rod, spring-loaded center point feeler, three straight feelers for IDs, and three curved feelers for ODs.
Catalog No. 50-005-27177 Supplier MSC Supplier No. 44509511
This coaxial centering indicator is designed to quickly center workpieces or fixtures on a mill. It operates at any angle from horizontal to vertical without changes, and can be used for both internal and external measurements by simply changing the feeler.
  • Quickly centers workpieces or fixtures on a mill.
  • Includes 6 feelers: 4 inch, 8 inch, 12 inch straight; 4 inch, 8 inch, 12 inch curved.
  • Operates at any angle from horizontal to vertical without requiring adjustments.
  • Adaptable for internal and external measurements by switching feelers.
  • Comes supplied in a fitted wood case.
  • Set includes indicator body, restraining rod, spring-loaded center point feeler, three straight feelers for IDs, and three curved feelers for ODs.