Learn More
MSC iGaging Electronic Caliper, 300 mm, 0.01 mm Resolution, IP54, Stainless Steel
Catalog No. 5000529289
Encompass
The electronic caliper is constructed from stainless steel and is designed for both inside and outside diameter measurements. It features an IP54 rating, providing protection against dust and splashing water. The caliper also includes a CR2032 lithium battery and offers data output capabilities compatible with SPC cables or wireless transmitters.
  • Constructed from durable stainless steel.
  • Measures both inside and outside diameters.
  • Features an IP54 rating for dust and splash protection.
  • Provides data output compatible with SPC cable or wireless transmitter.
  • Includes one CR2032 lithium battery for power.
  • Offers a measurement range up to 300 mm (12 in).
  • Delivers a resolution of ±0.01 mm (±0.0005 in).
  • Ensures an accuracy of ±0.03 mm (±0.0015 in).
  • Equipped with offset and nib jaws for versatile applications.
Catalog No. 50-005-29289 Supplier MSC Supplier No. 17621194
The electronic caliper is constructed from stainless steel and is designed for both inside and outside diameter measurements. It features an IP54 rating, providing protection against dust and splashing water. The caliper also includes a CR2032 lithium battery and offers data output capabilities compatible with SPC cables or wireless transmitters.
  • Constructed from durable stainless steel.
  • Measures both inside and outside diameters.
  • Features an IP54 rating for dust and splash protection.
  • Provides data output compatible with SPC cable or wireless transmitter.
  • Includes one CR2032 lithium battery for power.
  • Offers a measurement range up to 300 mm (12 in).
  • Delivers a resolution of ±0.01 mm (±0.0005 in).
  • Ensures an accuracy of ±0.03 mm (±0.0015 in).
  • Equipped with offset and nib jaws for versatile applications.