Learn More
MSC iGaging Electronic Caliper, 300 mm, 0.01 mm Resolution, IP67
Catalog No. 5000534838
Encompass
This electronic caliper offers precise measurements up to 300 mm (12 in) with a resolution of 0.01 mm (0.0005 in). Designed for various applications including outside, inside, step, and depth measurements, it features an IP67 rating for water and coolant resistance.
  • IP67 protective rating: water and coolant proof.
  • Ultra-response measure speed.
  • Inch/metric conversion.
  • High contrast LCD display.
  • 3V CR2032 battery included.
  • Supplied in protective case.
Catalog No. 50-005-34838 Supplier MSC Supplier No. 44504025
This electronic caliper offers precise measurements up to 300 mm (12 in) with a resolution of 0.01 mm (0.0005 in). Designed for various applications including outside, inside, step, and depth measurements, it features an IP67 rating for water and coolant resistance.
  • IP67 protective rating: water and coolant proof.
  • Ultra-response measure speed.
  • Inch/metric conversion.
  • High contrast LCD display.
  • 3V CR2032 battery included.
  • Supplied in protective case.