Learn More
MSC iGaging Electronic Caliper, 6 in, 0.0005 in Resolution, Stainless Steel
Catalog No. 5000529288
Encompass
This electronic caliper offers a measurement range of 0.0000in to 6in or 0mm to 150.00mm. It provides a resolution of ±0.0005in and accuracy of ±0.0010in. Constructed from stainless steel, this caliper is suitable for various measurements including inside diameter, step, outside diameter, and depth. It features a thumb wheel for adjustment and has an IP54 rating.
  • Stainless steel construction for both caliper and jaws.
  • Capable of measuring depth, inside diameter, outside diameter, and step.
  • IP54 rated for protection against dust and splashing water.
  • Includes inside diameter jaws and depth measurement rod.
  • Comes with a protective case.
  • Features a thumb wheel for fine adjustment.
  • Battery-powered with an included CR2032 lithium battery.
Catalog No. 50-005-29288 Supplier MSC Supplier No. 17620774
This electronic caliper offers a measurement range of 0.0000in to 6in or 0mm to 150.00mm. It provides a resolution of ±0.0005in and accuracy of ±0.0010in. Constructed from stainless steel, this caliper is suitable for various measurements including inside diameter, step, outside diameter, and depth. It features a thumb wheel for adjustment and has an IP54 rating.
  • Stainless steel construction for both caliper and jaws.
  • Capable of measuring depth, inside diameter, outside diameter, and step.
  • IP54 rated for protection against dust and splashing water.
  • Includes inside diameter jaws and depth measurement rod.
  • Comes with a protective case.
  • Features a thumb wheel for fine adjustment.
  • Battery-powered with an included CR2032 lithium battery.