Learn More
Cayman Chemical IgAmouse RabbIt Monclonal 50ug
SDP
Catalog No. 502947534
Encompass_Preferred
Immunogen: Mouse IgA • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgA; (-) Mouse IgG1, IgG2a, IgG2b, IgG2c, IgG3, IgM, IgE, (-) Human IgA • Applications: ELISA, WB (non-reducing conditions)
Catalog No. 50-294-7534 Supplier Cayman Chemical Supplier No. 3235550 UG
Immunogen: Mouse IgA • Host: Rabbit • Species Reactivity: (+) Mouse • Cross Reactivity: (+) IgA; (-) Mouse IgG1, IgG2a, IgG2b, IgG2c, IgG3, IgM, IgE, (-) Human IgA • Applications: ELISA, WB (non-reducing conditions)