Learn More
GENOVIS INC IgASAP Sub1 Lyophilized, 1000 Units, Lyophilized Enzyme for Above Hinge Digestion of Human IgA1
Catalog No. NC2739379
Encompass
IgASAP Sub1 is an IgA1-specific protease that digests human IgA1 at one specific site above the hinge, generating intact and homogeneous Fab and Fc fragments.
Catalog No. NC2739379 Supplier GENOVIS INC Supplier No. I0IA1010
IgASAP Sub1 is an IgA1-specific protease that digests human IgA1 at one specific site above the hinge, generating intact and homogeneous Fab and Fc fragments.