Learn More
Alpco Ltd IgD ELISA 96 Wells 5 L Research Use Only. Not for Use in Diagnostic Procedures.
SDP
Supplier:  Alpco Ltd 41IGDHUE01
Encompass
The IgD ELISA is intended for the quantitative measurement of IgD in human serum and plasma. - Results in just 1 hour and 20 minutes- Requires only 5 µL of sample Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3586