Learn More
Cayman Chemical IgDhumn RabbIt Monclonal 100ug
SDP
Catalog No. 502948439
Encompass_Preferred
Immunogen: Human IgD • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgD; (-) Human IgG, IgM, IgA, IgE • Applications: ELISA
Catalog No. 50-294-8439 Supplier Cayman Chemical Supplier No. 32117100 UG
Immunogen: Human IgD • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgD; (-) Human IgG, IgM, IgA, IgE • Applications: ELISA