Learn More
Cayman Chemical IgDhumn RabbIt Monclonal 50ug
SDP
Catalog No. 502950082
Encompass_Preferred
Immunogen: Human IgD • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgD; (-) Human IgG, IgM, IgE, IgA • Applications: ELISA
Catalog No. 50-295-0082 Supplier Cayman Chemical Supplier No. 3237150 UG
Immunogen: Human IgD • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgD; (-) Human IgG, IgM, IgE, IgA • Applications: ELISA