Learn More
Beckman Coulter Coulter Flow Cytometry, IgE, Single Color Antibody
Supplier:  Beckman Coulter A40174
Encompass
The Beckman Coulter A40174 Anti-Human IgE, FITC is a monoclonal antibody that binds to human IgE. It is conjugated to fluorescein isothiocyanate (FITC) and is intended for use in flow cytometry to identify and enumerate IgE-secreting cells. This antibody is supplied as a liquid in sodium azide. Recommended usage is 5-10 μL per test, with optimal dilution to be determined by titration. It can be used both alone and in combination with other antibodies. The antibody is designed for research use only.
Catalog No. NC1851527