Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgE Goat anti-Human, Biotin, Polyclonal , Abnova™

Catalog No. 89946194 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-946-194 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-946-194 Supplier Abnova Corporation Supplier No. PAB29740
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goat polyclonal antibody raised against human IgE.

Specifications

Antigen IgE
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Biotin
Description Goat polyclonal antibody raised against human IgE.
Dilution ELISA (0.01-0.1 ug/mL) Immunohistochemistry (0.2-2 ug/mL) Western Blot (0.01-0.1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.2% BSA, 0.09% sodium azide)
Host Species Goat
Immunogen Human IgE whole molecule.
Purification Method Antigen Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.