Learn More
Enzo Life Sciences IgE (human) monoclonal antibody (I27) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2270100
Encompass_Preferred
Clone: I27. Isotype: IgG1. Immunogen: Purified human IgE (preincubated with mouse serum anti human IgG). Species reactivity: Human. Specificity: Recognizes human IgE (heat labile epitope in N-terminal of CH3 domain). Applications: Flow Cytometry, WB, FUNC. Formulation: Liquid. In TRIS-NaCl, pH 8.
Catalog No. 50-201-2776