Learn More
Medchemexpress LLC IgE Human HEK293 50ug
SDP
Catalog No. 5000179352
Encompass_Preferred
IgE protein is an important immunoglobulin component that participates in the recognition and effector phases of humoral immunity As a membrane-bound receptor on B lymphocytes IgE triggers clonal expansion upon antigen binding leading to plasma cell differentiation IgE Protein Human (HEK293 His) is the recombinant human-derived IgE protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79352 Supplier Medchemexpress LLC Supplier No. HYP7390750UG
IgE protein is an important immunoglobulin component that participates in the recognition and effector phases of humoral immunity As a membrane-bound receptor on B lymphocytes IgE triggers clonal expansion upon antigen binding leading to plasma cell differentiation IgE Protein Human (HEK293 His) is the recombinant human-derived IgE protein expressed by HEK293 with C-His labeled tag