Learn More
Athens Research And Technology Inc IGE HUMAN PLASMA 100UG
Supplier:  Athens Research And Technology Inc 1616090705100UG
Encompass_Preferred
Immunoglobulin E, Human Plasma: 100 UG;Frozen in 100 mM Tris, pH 7.5, with 200 mM NaCl and 0.05% NaN3; MW 200,000; EC=1.33; Purity: > =95% by SDS-PAGE. This native protein is prepared and purified from human plasma in the Athens Research & Technology laboratory. Immunology, Molecular Biology Reagent
Catalog No. 50-489-781