Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgE Mouse anti-Human, Clone: 24A, Abnova™

Catalog No. 89952793 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-952-793 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-952-793 Supplier Abnova Corporation Supplier No. MAB12443
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human IgE.

Specifications

Antigen IgE
Applications ELISA, Western Blot
Classification Monoclonal
Clone 24A
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human IgE.
Dilution ELISA Western Blot The optimal working dilution should be determined by the end user.
Formulation Lyophilized
Host Species Mouse
Immunogen Human IgE whole molecule.
Purification Method Antigen Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity By indirect ELISA this antibody reacts with human IgE but not with human IgM, IgA or IgG.
Target Species Human
Content And Storage Store at 4°C. After reconstitution with 1mL of reagent quality water, aliquots should be stored at −20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.