Learn More
Medchemexpress LLC IgE Mouse HEK293 250ug | 250ug
SDP
Catalog No. 5000187435
Encompass_Preferred
IgE is an important component of the immunoglobulin chain that coordinates humoral immunity As a receptor membrane-bound IgE initiates clonal expansion and B cell differentiation IgE Protein Mouse (HEK293 His) is the recombinant mouse-derived IgE protein expressed by HEK293 with C-His C-6 His labeled tag
Catalog No. 50-001-87435 Supplier Medchemexpress LLC Supplier No. HYP77695250UG
IgE is an important component of the immunoglobulin chain that coordinates humoral immunity As a receptor membrane-bound IgE initiates clonal expansion and B cell differentiation IgE Protein Mouse (HEK293 His) is the recombinant mouse-derived IgE protein expressed by HEK293 with C-His C-6 His labeled tag