Learn More
Invitrogen™ Mouse IgE Uncoated ELISA Kit
Description
The Mouse Immunoglobulin E (IgE) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IgE. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Accession Number | P06336 |
| Assay Range | 4-250 ng/mL |
| Assay Sensitivity | 4 ng/mL |
| Content And Storage | Capture Antibody Concentrate Detection Antibody Concentrate mouse IgE isotype control (standard), lyophilized, 500 ng/mL upon reconstitution Phosphate Buffered Saline Concentrate (PBS) Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA) Pre-titrated Streptavidin-HRP Tetramethylbenzidine (TMB) Substrate Solution 96 Well Plate included with product Cat. Nos. ending in suffixes -22, -76, -86 |
| Conjugate | HRP |
| Immunoassay Kit Format | Uncoated ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.