Learn More
Enzo Life Sciences IgE (non-immune) (human) (1 mg)
SDP
Supplier:  Enzo Life Sciences BPDDIAHE11
Encompass_Preferred
Alternative name: Immunoglobulin E. Purity: ≥98% (SDS-PAGE). Formulation: Liquid. In 0.01M phosphate buffer, pH 7.4, containing 0.15M sodium chloride. Source: Purified human IgE with kappa light chains produced in vitro from a monoclonal hybridoma. Long Term Storage: . Applications: ICC, FUNC .
Catalog No. 50-201-2453