Learn More
Cayman Chemical IgEhumn RabbIt Monclonal 100ug
SDP
Catalog No. 502948191
Encompass_Preferred
Immunogen: Human IgE • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgE; (-) Human IgG, IgA, IgD, IgM • Application: ELISA
Catalog No. 50-294-8191 Supplier Cayman Chemical Supplier No. 32118100 UG
Immunogen: Human IgE • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) Human IgE; (-) Human IgG, IgA, IgD, IgM • Application: ELISA