Learn More
Cayman Chemical IgEhumn RabbIt Monclonal 50ug
SDP
Catalog No. 502946368
Encompass_Preferred
Immunogen: Human IgE • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgE; (-) Human IgG, IgM, IgD, IgA • Applications: ELISA
Catalog No. 50-294-6368 Supplier Cayman Chemical Supplier No. 3237250 UG
Immunogen: Human IgE • Host: Rabbit • Species Reactivity: (+) Human • Cross Reactivity: (+) IgE; (-) Human IgG, IgM, IgD, IgA • Applications: ELISA