Learn More
Sigma Aldrich Fine Chemicals Biosciences IGEPAL CA-630, 9002-93-1, MFCD00132505, 50 mL
Catalog No. NC1553470
Encompass
Linear Formula: (C2H4O)nC14H22O, for molecular biology, Synonym: Octylphenoxy poly(ethyleneoxy)ethanol, branched. IGEPAL CA-630 is a nonionic, non-denaturing detergent suitable for solubilization, isolation and purification of membrane protein complexes.It is chemically indistinguishable from Nonidet P-40.
Catalog No. NC1553470 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. I889650ML
Linear Formula: (C2H4O)nC14H22O, for molecular biology, Synonym: Octylphenoxy poly(ethyleneoxy)ethanol, branched. IGEPAL CA-630 is a nonionic, non-denaturing detergent suitable for solubilization, isolation and purification of membrane protein complexes.It is chemically indistinguishable from Nonidet P-40.