Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich IGEPAL™ CO-520 average Mn 441

Catalog No. 238643100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-864-3100G 100 g
23-864-3500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-864-3100G Supplier Sigma Aldrich Supplier No. 238643100G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

IGEPAL(R)CO-520 or poly (oxyethylene)nonylphenyl ether) is a non-ionic surfactant.Igepal aggregates have been used in fluorescence and light-scattering studies.Used in the preparation of monodispersed colloidal silica cadmium suphide nanocomposites, alumina nanoparticles, ultra thin silica coated magnetic nanoparticles.IGEPAL is a registered trademark of Solvay

Specifications

CAS 68412-54-4
Density 0.997 g/mL (at 25°C)
Linear Formula (C2H4O)n -+ C15H24O, n ∼5
Refractive Index n20/D 1.495 (literature)
MDL Number MFCD00132851
Quantity 100 g
Synonym Polyoxyethylene (5) nonylphenylether, branched
Molecular Weight (g/mol) 441
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.