Learn More
Abcam IGEPAL CA-630 MegaPure™ Detergent,10% Solution, 100ML
Catalog No. 502215527
Encompass_Preferred
MW 514.7 Da. A 10% solution of the nonionic surfactant IGEPAL CA-630 in water. Specially processed to remove the aldehydes and peroxides. Lacks the UV absorbance that causes interference in spectophotometric monitoring and assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5527 Supplier Abcam Supplier No. AB285400100ML
MW 514.7 Da. A 10% solution of the nonionic surfactant IGEPAL CA-630 in water. Specially processed to remove the aldehydes and peroxides. Lacks the UV absorbance that causes interference in spectophotometric monitoring and assays.
The product is subject to the following: Abcam Restricted Use Statement