Learn More
Crystal Chem Inc IGF-1 ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80572
Encompass
The IGF-1 ELISA is used to quantify levels of human IGF-1 in plasma, serum, and cell culture using only a 5 µL sample volume. Highlights: Kits use only 10 µL sample. Dynamic range of 2 - 50 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human research markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7739