Learn More
Alpco Ltd Mouse / Rat IGF-1 ELISA
SDP
Supplier:  Alpco Ltd 22IG1MSE01
Encompass
The Mouse / Rat IGF-1 ELISA is for the quantitative determination of IGF-1 in rat and mouse serum and plasma (EDTA-, Citrate-, and Heparin-).
Dynamic range
0.5-18 ng/mL
Sensitivity
0.315 ng/mL
Sample size
5 µL
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-921-879