Learn More
Cayman Chemical IGF-1 (human, recombinant), Purity: ≥95% estimated by SDS-PAGE, 1 mg
SDP
Catalog No. 502948579
Encompass_Preferred
Source: Active recombinant human IGF-1 expressed in E. coli • Amino acids: Met1; 49-118 • MW: 7.8 kDa
Catalog No. 50-294-8579 Supplier Cayman Chemical Supplier No. 339811 MG
Source: Active recombinant human IGF-1 expressed in E. coli • Amino acids: Met1; 49-118 • MW: 7.8 kDa