Learn More
Eurofins Discoverx IGF-1R, activated, 10 µg. (14-802)
Supplier:  Eurofins Discoverx 14802
Encompass
N-terminal 6xHis-tagged recombinant, human IGF-1R amino acids 959-end. Auto activated on column by incubating with Mg/ATP, excess ATP and MgAc removed by multiple column wash steps. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9359