Learn More
Enzo Life Sciences IGF 1Rtide (1mg)
SDP
Catalog No. 502011317
Encompass_Preferred
This peptide, KKKSPGEYVNIEFG, is derived from human IRS-1 and is a substrate for TRK1, JAK2, and RET kinases. Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1317 Supplier Enzo Life Sciences Supplier No. BMLP2570001
This peptide, KKKSPGEYVNIEFG, is derived from human IRS-1 and is a substrate for TRK1, JAK2, and RET kinases. Purity: ≥95%. Long Term Storage: -20°C.