Learn More
Medchemexpress LLC HY-P7019 100ug Medchemexpress, IGF-2, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP7019100UG
Encompass_Preferred
Medchemexpress, HY-P7019 100ug IGF-2, Human CAS: Recombinant Human Insulin-like Growth factor-2 is a mitogenic cytokine, binds to IGF type 1 receptor, and modulates growth in many tissues, such as nervous tissue, lymphoid tissue, reproductive tissue, smooth muscle, endothelium, and bone. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4272