Learn More
Genscript Corporation Antibody Protein Proteins Proteins 1 MG 1/EA
Supplier:  Genscript Corporation Z032011
Encompass
IGF-BP-2 also known as Insulin-like growth factor-binding protein 2 IBP-2 and BP-2 is a cysteine-rich secreted protein belonging to the IGF-binding protein superfamily. It is expressed by the central nervous system bone cells and reproductive tissues. IGF-BP-2 binds to both IGF-I and IGF-II with a much higher binding affinity to IGF-II than IGF-I. IGF-BP-2 has been shown to inhibitand stimulate the growth promoting effects of IGFs thus serving as a regulator for IGF distribution function and activity.
Catalog No. 50-252-9727