Learn More
Medchemexpress LLC HY-P7021 10ug Medchemexpress, IGF-BP5, Human CAS: Purity:>98%
SDP
Catalog No. 501970554
Encompass_Preferred
Medchemexpress, HY-P7021 10ug IGF-BP5, Human CAS: Recombinant Human Insulin-like Growth Factor-Binding Protein 5 is a unique insulin-like growth factor-binding protein, modulates the actions of secreted insulin-like growth factors, and stimulates osteoblast proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0554 Supplier Medchemexpress LLC Supplier No. HYP702110UG
Medchemexpress, HY-P7021 10ug IGF-BP5, Human CAS: Recombinant Human Insulin-like Growth Factor-Binding Protein 5 is a unique insulin-like growth factor-binding protein, modulates the actions of secreted insulin-like growth factors, and stimulates osteoblast proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.