Learn More
CPC Scientific H-Gly-Tyr-Gly-Ser-Ser-Ser-Arg-Arg-Ala-Pro-Gln-Thr-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109588
Encompass
The IGF fragment has been synthesized to generate an antibody to the C-peptide region of IGF-I.ONE-LETTER SEQUENCE: GYGSSSRRAPQTMOLECULAR FORMULA: C51H83N19O19MOLECULAR WEIGHT:1266.34STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [82177-09-1], RESEARCH AREA: DiabetesREFERENCES: R.L.Hintz et al., J. Clin. Endocrinol. Metab., 50, 405 (1980)
Catalog No. 50-210-9588 Supplier CPC Scientific Supplier No. ILGF004A
The IGF fragment has been synthesized to generate an antibody to the C-peptide region of IGF-I.ONE-LETTER SEQUENCE: GYGSSSRRAPQTMOLECULAR FORMULA: C51H83N19O19MOLECULAR WEIGHT:1266.34STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [82177-09-1], RESEARCH AREA: DiabetesREFERENCES: R.L.Hintz et al., J. Clin. Endocrinol. Metab., 50, 405 (1980)