Learn More
Medchemexpress LLC HY-P1777 1mg Medchemexpress, IGF-I (24-41) CAS:135861-49-3 Purity:>98%
Supplier: Medchemexpress LLC HYP17771MG
Medchemexpress, HY-P1777 1mg IGF-I (24-41) CAS:135861-49-3 IGF-I (24-41) is amino acids 24 to 41 fragment of Insulin-like Growth Factor I (IGF-I). IGF-I is partly responsible for systemic GH activities although it possesses a wide number of own properties (anabolic, antioxidant, anti-inflammatory and cytoprotective actions). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.