Learn More
Medchemexpress LLC IGF-I (24-41) (Insulin-like Growth Factor I (24-41)) | 135861-49-3 | 99.1% | 2017.16 | 5 MG
SDP
Catalog No. 5000422604
Encompass_Preferred
IGF-I (24-41) (Insulin-like Growth Factor I (24-41)) is a fragment of IGF-I, specifically amino acids 24 to 41. IGF-I is a 70 amino acid polypeptide hormone that acts as a trophic factor for both neurons and glial cells and plays a role in systemic growth hormone (GH) activities. It exhibits anabolic, antioxidant, anti-inflammatory, and cytoprotective actions and regulates somatic growth and behavioral development.
  • Acts as a trophic factor for neurons and glial cells
  • Exhibits anabolic, antioxidant, anti-inflammatory, and cytoprotective actions
  • Regulates somatic growth and behavioral development
  • For research use only
Catalog No. 50-004-22604 Supplier Medchemexpress LLC Supplier No. HYP17775MG
Add to Cart
Add to Cart
IGF-I (24-41) (Insulin-like Growth Factor I (24-41)) is a fragment of IGF-I, specifically amino acids 24 to 41. IGF-I is a 70 amino acid polypeptide hormone that acts as a trophic factor for both neurons and glial cells and plays a role in systemic growth hormone (GH) activities. It exhibits anabolic, antioxidant, anti-inflammatory, and cytoprotective actions and regulates somatic growth and behavioral development.
  • Acts as a trophic factor for neurons and glial cells
  • Exhibits anabolic, antioxidant, anti-inflammatory, and cytoprotective actions
  • Regulates somatic growth and behavioral development
  • For research use only