Learn More
CPC Scientific H-Gly-Pro-Glu-OH (trifluoroacetate salt) 250MG
SDP
Supplier:  CPC Scientific ILGF003A
Encompass
This N-terminal tripeptide of IGF-I facilitated the in vitro release of acetylcholine with a several hundredfold higher potency (10-10 - 10-6 M) than intact IGF-I (4 ? 10-8 M), whereas truncated IGF-I lacking the tripeptide GPE, did not show any significant effect. This raises the possibility that GPE may be the active site of IGF-I. - Although the precise mode of action of GPE is still unknown, another study suggests that local administration of GPE is neuroprotective after brain HI injury via glial cells. In addition, systemic administration of GPE showed a more widespread neuroprotective effect. Therefore, GPE may represent a complementary pathway for central and systemic IGF-1's antiapoptotic effects.SEQUENCE: H-Gly-Pro-Glu-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: GPEMOLECULAR FORMULA: C12H19N3O6MOLECULAR WEIGHT: 301.3STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [32302-76-4]SYNONYMS: RESEARCH AREA: DiabetesREFERENCES:L.Nilsson-Hakansson et al., Neuro
Catalog No. 50-210-9587