Learn More
Bachem IGF-I (24-41) Trifluoroacetate
SDP
Supplier:  Bachem 4030780.0005
Encompass_Preferred
IGF-I (24-41) Trifluoroacetate, H-Tyr-Phe-Asn-Lys-Pro-Thr-Gly-Tyr-Gly-Ser-Ser-Ser-Arg-Arg-Ala-Pro-Gln-Thr-OH, 135861-49-3, C88H133N27O28, Mr 2017.19, 5mg. Insulin-Like Growth Factor I (24-41). IGF-I and IGF-I (24-41) affected somatic and neurobehavioral development of newborn male mice.
Catalog No. 50-194-6904