Learn More
Alpco Ltd IGF-1 ELISA
SDP
Supplier:  Alpco Ltd 22IGFHUE01
Encompass
The IGF-1 ELISA is for the quantitative determination of IGF-1 in human serum and plasma.
Dynamic range
2-50 ng/mL
Sensitivity
0.09 ng/mL
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3538