Learn More
Ansh Labs IGF-I ELISA 96-wells, 50 L Plasma/Serum, Range 6 calibrators ranging 0.48-32.2 ng/mL, LOD 0.025 ng/mL, Time 1 hour, Shelf Life 24 months
SDP
Supplier:  Ansh Labs AL121
Encompass
IGF-I ELISA has a total 1 hour incubation, calibrators and range of 6, 0.48-32.2 ng/mL, sensitivity of 0.025 ng/mL, sample of 50 µL Plasma, Serum. Reacts with Human. Crossreactivity with related molecules is not significant.Ansh Labs is a USA FDA registered manufacturer of diagnostic test kits, antibodies, antigens, and proteins.
Catalog No. 50-194-3730