Learn More
BioVendor, LLC Insulin Like Growth Factor-1 (IGF-I) Human ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC E20
Encompass
Insulin Like Growth Factor-1 (IGF-I) Human ELISA (96 wells). Research Use Only. Immunoassay kit for quantification of human IGF1 in serum or plasma samples. Quality control sera included. CALIBRATION RANGE: 150 ng/ml. LIMIT OF DETECTION: 0.09 ng/ml. BIOMARKER SUMMARY: Due to its GH-dependence, determination of serum IGF-I was shown to be a useful tool in diagnosis of growth disorders, especially with regard to GH deficiency (GHD) or acromegaly. The major advantage of IGFI determination compared to GH determination is its stable circadian concentration; therefore a single measurement is sufficient. Hence IGFI determination should be the first in a series of laboratory test. Clearly normal levels would then rule out disturbances of the GH-IGF-I-axis.
Catalog No. 50-206-9070