Learn More
Medchemexpress LLC IGF-I/IGF-1 Protein, Mouse | 96.7% | Approximately 8 kDa | 250 UG
SDP
Catalog No. 5000396589
Encompass_Preferred
IGF-I/IGF-1 Protein is an insulin-like growth factor involved in growth-promoting, metabolic-regulating, neuroprotective, and neuroplasticity-regulating activities. This recombinant protein is expressed by E. coli without a tag and plays a key role in cell growth, differentiation, survival, and cell cycle regulation.
  • Possesses growth-promoting and metabolic-regulating functions
  • Exhibits neuroprotective and neuroplasticity-regulating activities
  • Key role in growth and development, cell proliferation, and metabolic regulation
  • Expressed by E. coli without a tag
  • Stimulates osteoblast proliferation and differentiation
  • Promotes neuronal survival
  • Promotes proliferation of thyroid epithelial cells at 1 ng/mL for 6 h
  • ED50 is less than 10 ng/mL in FDC-P1 cells
Catalog No. 50-003-96589 Supplier Medchemexpress LLC Supplier No. 250UGHYP7070
IGF-I/IGF-1 Protein is an insulin-like growth factor involved in growth-promoting, metabolic-regulating, neuroprotective, and neuroplasticity-regulating activities. This recombinant protein is expressed by E. coli without a tag and plays a key role in cell growth, differentiation, survival, and cell cycle regulation.
  • Possesses growth-promoting and metabolic-regulating functions
  • Exhibits neuroprotective and neuroplasticity-regulating activities
  • Key role in growth and development, cell proliferation, and metabolic regulation
  • Expressed by E. coli without a tag
  • Stimulates osteoblast proliferation and differentiation
  • Promotes neuronal survival
  • Promotes proliferation of thyroid epithelial cells at 1 ng/mL for 6 h
  • ED50 is less than 10 ng/mL in FDC-P1 cells