Learn More
Genscript Corporation Antibody Protein Proteins Proteins 50 UG 1/EA
Supplier:  Genscript Corporation Z0317850
Encompass
Insulin-like Growth Factor I (IGF-I) is a single chain 7 kDa growth-promoting polypeptide originally identified as somatomedin-c. It belongs to the IGF family of peptides which also includes IGF-II and insulin. The gene expression of IGF-I is mainly regulated by Growth Hormone and IGF-I executes its functions via signaling through transmembrane tyrosine receptors (IGF Receptors). Most circulating IFG-I is associated with the IGF Binding Protein 3 (IGFBP-3) and the IGFBPs may inhibit the actions of IGFs by competing against the IGF Receptors. IGF-I is active in post-natal and adult animals and is crucial for somatic growth as IGF-I null mice show marked retardation in utero. IGF-I is involved in carcinogenesis and related to prostate cancer as well.
Catalog No. 50-252-9738