Learn More
Medchemexpress LLC IGF-I R Human HEK2 10ug
SDP
Catalog No. 5000181065
Encompass_Preferred
IGF-I receptor (IGF1R) mediates insulin-like growth factor effects binding strongly to IGF1 Activation triggers PI3K-AKT/PKB and Ras-MAPK pathways influencing cell survival protein synthesis and proliferation IGF1R also signals through JAK/STAT inhibiting JNK activation Hybrid receptors show variable binding to IGF1 and insulin IGF-I R Protein Human (HEK293 His) is the recombinant human-derived IGF-I R protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-81065 Supplier Medchemexpress LLC Supplier No. HYP7260610UG
IGF-I receptor (IGF1R) mediates insulin-like growth factor effects binding strongly to IGF1 Activation triggers PI3K-AKT/PKB and Ras-MAPK pathways influencing cell survival protein synthesis and proliferation IGF1R also signals through JAK/STAT inhibiting JNK activation Hybrid receptors show variable binding to IGF1 and insulin IGF-I R Protein Human (HEK293 His) is the recombinant human-derived IGF-I R protein expressed by HEK293 with C-6 His labeled tag