Learn More
Medchemexpress LLC IGF-I R Rat HEK293 2ug | 2ug
SDP
Catalog No. 5000194336
Encompass_Preferred
Igf1r Protein a cell surface receptor is involved in growth regulation and development Dysregulation of Igf1r Protein has been linked to diseases like cancer and growth disorders Targeting Igf1r Protein may offer potential therapeutic interventions in these conditions by modulating cell growth inhibiting tumor progression and managing growth-related disorders IGF-I R Protein Rat (HEK293 His) is the recombinant rat-derived IGF-I R protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-94336 Supplier Medchemexpress LLC Supplier No. HYP787242UG
Igf1r Protein a cell surface receptor is involved in growth regulation and development Dysregulation of Igf1r Protein has been linked to diseases like cancer and growth disorders Targeting Igf1r Protein may offer potential therapeutic interventions in these conditions by modulating cell growth inhibiting tumor progression and managing growth-related disorders IGF-I R Protein Rat (HEK293 His) is the recombinant rat-derived IGF-I R protein expressed by HEK293 with C-His labeled tag