Learn More
Genscript Corporation Antibody Protein Proteins Proteins 100 UG 1/EA
Supplier:  Genscript Corporation Z03726100
Encompass
Insulin-like growth factor 1 (IGF-1) also called Somatomedin is a hormone similar in molecular structure to insulin but has a much higher growth-promoting activity. IGF-1 consists of 70 amino acids in a single chain with three intramolecular disulfide bridges. IGF-1 may be a physiological regulator of [1-14C]-2-deoxy-D-glucose (5DG) transport and glycogen synthesis in osteoblasts. It is able to stimulate glucose transport in bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin not only regarding glycogen and DNA synthesis but also with regard to enhancing glucose uptake. It may also play a role in synapse maturation.
Catalog No. 50-252-9571