Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IGF-II Protein, Avitag™,Fc Tag
Catalog No. 501957538
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IGF-II, Avitag,Fc Tag (IG2-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession # P01344-1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 35.3 kDa. As a result of glycosylation, the protein migrates as 40 kDa under reducing (R) condition, and 66 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human IGFBP-3, His Tag (Cat. No. IG3-H5229) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IGF-II, Avitag,Fc Tag (Cat. No. IG2-H82F9) with a linear range of 0.008-0.1 μg/mL (QC tested).
Catalog No. 50-195-7538 Supplier ACROBiosystems Supplier No. IG2H82F925UG
Recombinant Protein;25UG;Biotinylated Human IGF-II, Avitag,Fc Tag (IG2-H82F9) is expressed from human 293 cells (HEK293). It contains AA Ala 25 - Glu 91 (Accession # P01344-1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 35.3 kDa. As a result of glycosylation, the protein migrates as 40 kDa under reducing (R) condition, and 66 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human IGFBP-3, His Tag (Cat. No. IG3-H5229) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IGF-II, Avitag,Fc Tag (Cat. No. IG2-H82F9) with a linear range of 0.008-0.1 μg/mL (QC tested).