Learn More
Carnabio Usa Inc IGF1R/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 081415
Encompass
IGF1R, Cytoplasmic domain, Wild type, Amino Acid 959-1367, NP_000866.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0463