Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IGF1R, His Tag Recombinant Protein

Catalog No. PR4654A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR4654A 100 μg
PV3250 10 μg
PV3592 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PR4654A Supplier Invitrogen™ Supplier No. PR4654A
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic.

IGF1R, also known as Insulin-like Growth Factor 1 (IGF-1) Receptor, is a receptor tyrosine kinase activated by IGF-1 and IGF-2. IGFR has been implicated in a variety of cancers. The protein is a tetramer consisting of two alpha, and two beta subunits. The alpha-subunits binds to insulin growth factor 1 with higher affinity than to insulin growth factor 2. Binding results in a conformational change, followed by autophosphorylation of IGF-1R at positions 1134, 1135, 1136. The activated IGF-1R in turn, causes phosphorylation of substrate proteins, followed by sequential activation of RAS, RAF, and mitogen-activated protein kinase isoforms ERK, p38, and JNK, leading to the transcription of genes that drive proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08069
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 100mM NaCl, 2mM DTT, 0.05% Triton X-100, 50% glycerol, 0.05mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 3480
Molecular Weight (g/mol) 48.9 kDa
Name Human IGF1R, His Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias A330103N21Rik; CD221; CD221 antigen; D930020L01; hyft; IGF 1 receptor; IGF 1R; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; Insulin like growth factor 1 receptor precursor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2
Common Name IGF1R (CD221)
Gene Symbol IGF1R
Kinase Family Insulin & IGF Receptor Kinase Family
Protein Length 960-1397
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence 960-1397
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Tyrosine Kinases
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic Domain, Cytoplasmic
Form Liquid
Protein Subtype Receptor Tyrosine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.