Learn More
Sino Biological Peptide Substrates: IGF1Rtide
Catalog No. 502568948
Encompass_Preferred
The 14 amino acids of IGF1Rtide peptide sequence (KKKSPGEYVNIEFG) is derived from human IRS-1 protein residues 891-902.
Catalog No. 50-256-8948 Supplier Sino Biological Supplier No. I1558
The 14 amino acids of IGF1Rtide peptide sequence (KKKSPGEYVNIEFG) is derived from human IRS-1 protein residues 891-902.