Learn More
Aviva Systems Biology Corp IGF2 Antibody - C-terminal region (ARP54342_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54342_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IGF2. It was validated on Western Blot. . Alternate names: C11orf43, FLJ22066, FLJ44734, INSIGF, pp9974, IGF-II, PP9974ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-285-908